4-Butoxybenzyl alcohol ≥97.0%
Supplier: TCI America
B1042-5G
B1042-25G
TCB1042-005GEA
52.93
CAD
TCB1042-005G
TCB1042-025G
4-Butoxybenzyl alcohol ≥97.0%
4-Butoxybenzyl alcohol
CAS Number: 6214-45-5
MDL Number: MFCD00004657
Molecular Formula: C11H16O2
Molecular Weight: 180.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30
Formula:
C₁₁H₁₆O₂ Boiling Pt: 169…171 °C (22 mmHg) Melting Pt: 32…32.5 °C Flash Pt: 110 °C |
MDL Number:
MFCD00004657 CAS Number: 6214-45-5 EINECS: 228-284-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...