Bistrifluoroacetamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

B0986-25G B0986-5G
TCB0986-025GEA 808.33 CAD
TCB0986-025G TCB0986-5G
Bistrifluoroacetamide ≥98.0% (by total nitrogen basis)
Bistrifluoroacetamide

CAS Number: 407-24-9
MDL Number: MFCD00013564
Molecular Formula: C4HF6NO2
Molecular Weight: 209.05
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 84


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: BTFA

Formula: C₄HF₆NO₂
Boiling Pt: 120 °C
Melting Pt: 84 °C
MDL Number: MFCD00013564
CAS Number: 407-24-9
EINECS: 206-981-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbd217a985e61076cb66fbf4b78fcbc93977800d&vid=bbd217a985e61076cb66fbf4b78fcbc93977800d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR