Benzenepentacarboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B0952-5G
B0952-25G
TCB0952-005GEA
1042.8
CAD
TCB0952-005G
TCB0952-25G
Benzenepentacarboxylic acid ≥98.0% (by HPLC, titration analysis)
Benzenepentacarboxylic acid
CAS Number: 1585-40-6
MDL Number: MFCD00020281
Molecular Formula: C11H6O10
Molecular Weight: 298.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₁₁H₆O₁₀ |
MDL Number:
MFCD00020281 CAS Number: 1585-40-6 EINECS: 216-441-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...