1-Bromoheptadecane ≥98.0%

Supplier: TCI America

B0937-5G B0937-25G
TCB0937-005GEA 247.59 CAD
TCB0937-005G TCB0937-025G
1-Bromoheptadecane ≥98.0%
1-Bromoheptadecane

CAS Number: 3508-00-7
MDL Number: MFCD00017891
Molecular Formula: C17H35Br
Molecular Weight: 319.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 29


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Heptadecyl Bromide

Formula: BrH₂C(CH₂)₁₅CH₃
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 32 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017891
CAS Number: 3508-00-7
EINECS: 222-506-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=907ce936cca24f39e1f865dac8b83ef73ca36604&vid=70ea2f2737522e1280ffdad0776a69dca96e5713&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR