2-Butyl-2-ethyl-1,3-propanediol ≥98.0%
Supplier: TCI America
Synonyms:
2-Ethyl-2-(hydroxymethyl)-1-hexanol, 3,3-Bis(hydroxymethyl)heptane, 2-Butyl-2-ethylpropane-1,3-diol
CAS Number: 115-84-4
MDL Number: MFCD00004697
Molecular Formula: C9H20O2
Molecular Weight: 160.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 43
Flash Point (°C): 110
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,3-Bis(hydroxymethyl)heptane, 2-Ethyl-2-(hydroxymethyl)-1-hexanol
Formula:
C₉H₂₀O₂ Boiling Pt: 178 °C Melting Pt: 43 °C Density: 0.931 g/cm³ (50 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004697 CAS Number: 115-84-4 EINECS: 204-111-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...