1,2,3-Butanetriol ≥90.0%

Supplier: TCI America

B0918-25ML B0918-5ML
TCB0918-025MLEA 2592.78 CAD
TCB0918-025ML TCB0918-005ML
1,2,3-Butanetriol ≥90.0%
1,2,3-Butanetriol

CAS Number: 4435-50-1
MDL Number: MFCD00059662
Molecular Formula: C4H10O3
Molecular Weight: 106.12
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 1,2,3-Trihydroxybutane

Formula: C₄H₁₀O₃
Boiling Pt: 175 °C
Density: 1.18 (20 °C)
MDL Number: MFCD00059662
CAS Number: 4435-50-1
EINECS: 224-643-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ea9885bb3de121800a74fdd7233f1981929ebfc&vid=8ea9885bb3de121800a74fdd7233f1981929ebfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR