2,4'-Bipyridyl ≥97.0%

Supplier: TCI America

B0863-5G B0863-1G
TCB0863-5GEA 282.82 CAD
TCB0863-5G TCB0863-001G
2,4'-Bipyridyl ≥97.0%
2,4'-Bipyridyl

CAS Number: 581-47-5
MDL Number: MFCD00006217
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 64


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 2,4'-Bipyridine, 2,4'-Dipyridyl

Formula: C₁₀H₈N₂
Boiling Pt: 110 °C
Melting Pt: 64 °C
MDL Number: MFCD00006217
CAS Number: 581-47-5
EINECS: 209-467-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec5d2d51284926ee999a956b568c2862eae4b55e&vid=ec5d2d51284926ee999a956b568c2862eae4b55e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR