4-Bromoresorcinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0654-5G B0654-25G
TCB0654-005GEA 209.98 CAD
TCB0654-005G TCB0654-025G
4-Bromoresorcinol ≥98.0% (by GC, titration analysis)
4-Bromoresorcinol

CAS Number: 6626-15-9
MDL Number: MFCD00002272
Molecular Formula: C6H5BrO2
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 103

Formula: C₆H₅BrO₂
Boiling Pt: 150 °C
Melting Pt: 103 °C
MDL Number: MFCD00002272
CAS Number: 6626-15-9
EINECS: 229-586-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b72eba01370036b9e0d5523f6098ac20de542f27&vid=b72eba01370036b9e0d5523f6098ac20de542f27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR