Bromophenol red ≥80.0% (by HPLC)

Supplier: TCI America

B0632-1G B0632-25G
TCB0632-001GEA 99.54 CAD
TCB0632-001G TCB0632-025G
Bromophenol red ≥80.0% (by HPLC)
Bromophenol red

CAS Number: 2800-80-8
MDL Number: MFCD00023014
Molecular Formula: C19H12Br2O5S
Molecular Weight: 512.17
Purity/Analysis Method: >80.0% (HPLC)
Form: Crystal
Lambda max.: 576 nm (pH 6.8 buffer sol.)

Formula: C₁₉H₁₂Br₂O₅S
Melting Pt: 230 °C
MDL Number: MFCD00023014
CAS Number: 2800-80-8
EINECS: 220-538-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fdac9ed5a95402f60eab62051431bd599c30edc&vid=9fdac9ed5a95402f60eab62051431bd599c30edc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR