1-Bromonaphthalene ≥95.0%

Supplier: TCI America
Warning

Synonyms: α-Bromonaphthalene

B0618-25G B0618-500G B0618-100G
TCB0618-025GEA 25.72 CAD
TCB0618-025G TCB0618-500G TCB0618-100G
1-Bromonaphthalene ≥95.0%
1-Bromonaphthalene

CAS Number: 90-11-9
MDL Number: MFCD00003868
Molecular Formula: C10H7Br
Molecular Weight: 207.07
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 282
Melting point (°C): -2
Flash Point (°C): 110
Specific Gravity (20/20): 1.48

Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 282 °C
Melting Pt: –2 °C
Density: 1.48 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003868
CAS Number: 90-11-9
EINECS: 201-965-2
Merck Index: 13,01410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f643600463eb577931a1aa8e69f6360344d817c4&vid=f643600463eb577931a1aa8e69f6360344d817c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR