Xylenol orange disodium salt

Supplier: TCI America

Synonyms: 3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulphonephthalein disodium salt, 3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt

B0477-1G B0477-25G B0477-5G
TCB0477-001GEA 120.59 CAD
TCB0477-001G TCB0477-025G TCB0477-5G
Xylenol orange disodium salt
Xylenol orange disodium salt

CAS Number: 1611-35-4
MDL Number: MFCD00014580
Molecular Formula: C31H32N2O13S
Molecular Weight: 716.62
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: 3,3'-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonphthalein Disodium Salt

Formula: C₃₁H₃₀N₂Na₂O₁₃S
MW: 716.62 g/mol
MDL Number: MFCD00014580
CAS Number: 1202864-39-8
EINECS: 216-553-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c03571a71b788b6848906dbf8c6e81eaf372729&vid=1c03571a71b788b6848906dbf8c6e81eaf372729&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR