Benzylmalonic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Benzylmalonic acid

B0422-5G B0422-25G
TCB0422-005GEA 98.07 CAD
TCB0422-005G TCB0422-025G
Benzylmalonic acid ≥98.0%
Benzylmalonic acid

CAS Number: 616-75-1
MDL Number: MFCD00004261
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118

Formula: C₁₀H₁₀O₄
Melting Pt: 118 °C
MDL Number: MFCD00004261
CAS Number: 616-75-1
EINECS: 210-491-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b38bbf7c231f0277688bf8a3e65b2dd799ef74e7&vid=b38bbf7c231f0277688bf8a3e65b2dd799ef74e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR