1-Bromodocosane ≥98.0%

Supplier: TCI America
Warning

B0392-25G B0392-250G
TCB0392-025GEA 144.14 CAD
TCB0392-025G TCB0392-250G
1-Bromodocosane ≥98.0%
1-Bromodocosane

CAS Number: 6938-66-5
MDL Number: MFCD00013543
Molecular Formula: C22H45Br
Molecular Weight: 389.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Docosyl Bromide

Formula: BrH₂C(CH₂)₂₀CH₃
Melting Pt: 44 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013543
CAS Number: 6938-66-5
EINECS: 230-069-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36b1df13a1392e6e8b3f084332d313e5b1af7b09&vid=e9d692ba56efff156caa669f9bb4548986a2f6bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR