11-Bromoundecanoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B0389-25G B0389-500G B0389-100G
TCB0389-025GEA 71.1 CAD
TCB0389-025G TCB0389-500G TCB0389-100G
11-Bromoundecanoic acid ≥97.0% (by GC, titration analysis)
11-Bromoundecanoic acid

CAS Number: 2834-05-1
MDL Number: MFCD00002732
Molecular Formula: C11H21BrO2
Molecular Weight: 265.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: 11-Bromohendecanoic Acid

Formula: BrH₂C(CH₂)₉COOH
Boiling Pt: 190 °C
Melting Pt: 49 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002732
CAS Number: 2834-05-1
EINECS: 220-602-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7238cd6fde65b454bf44b0e59ba2f8ee6f62d4e&vid=f7238cd6fde65b454bf44b0e59ba2f8ee6f62d4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR