2-Benzoylpyridine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B0304-25G B0304-500G
TCB0304-025GEA 28.95 CAD
TCB0304-025G TCB0304-500G
2-Benzoylpyridine ≥99.0% (by GC, titration analysis)
2-Benzoylpyridine

CAS Number: 91-02-1
MDL Number: MFCD00006300
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 167


Other Product Names / Synonyms: Phenyl 2-Pyridyl Ketone

Formula: C₁₂H₉NO
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 44 °C
Density: 1.171 g/cm³ (0 °C)
Flash Pt: 167 °C
MDL Number: MFCD00006300
CAS Number: 91-02-1
EINECS: 202-034-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8295a0b2dce9faa8904142c0d7c5d1e10e16afe3&vid=8295a0b2dce9faa8904142c0d7c5d1e10e16afe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR