Mellitic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0246-25G B0246-5G
TCB0246-25GEA 1068.08 CAD
TCB0246-25G TCB0246-005G
Mellitic acid ≥98.0% (by titrimetric analysis)
Mellitic acid

CAS Number: 517-60-2
MDL Number: MFCD00002469
Molecular Formula: C12H6O12
Molecular Weight: 342.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 288


Other Product Names / Synonyms: Benzenehexacarboxylic Acid

Formula: C₁₂H₆O₁₂
Melting Pt: 288 °C
Storage Temperature: Ambient
MDL Number: MFCD00002469
CAS Number: 517-60-2
EINECS: 208-243-6
Merck Index: 12,05868
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd5d5adec4c5af375cc5098d9d40201dde1b2ee8&vid=4bf96a8a135e269efbdb82ca52d620a924180737&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR