Propyl benzoate ≥99.0%

Supplier: TCI America
Warning

B0076-25ML B0076-500ML
TCB0076-025MLEA 45.82 CAD
TCB0076-025ML TCB0076-500ML
Propyl benzoate ≥99.0%
Propyl benzoate

CAS Number: 2315-68-6
MDL Number: MFCD00009370
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 112
Freezing point (°C): -52
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Benzoic Acid Propyl Ester

Formula: C₁₀H₁₂O₂
Boiling Pt: 230 °C
Melting Pt: –52 °C
Density: 1.02 (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009370
CAS Number: 2315-68-6
EINECS: 219-020-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50e89f2890038cdb6d5e4d7842a679572ffbe4ca&vid=143bfefdaae5513875ef6aa2ad60e3eff2ca971d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR