Pyromellitic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0039-25G B0039-500G B0039-100G
TCB0039-025GEA 34.98 CAD
TCB0039-025G TCB0039-500G TCB0039-100G
Pyromellitic acid ≥98.0% (by titrimetric analysis)
Pyromellitic acid

CAS Number: 89-05-4
MDL Number: MFCD00002471
Molecular Formula: C10H6O8
Molecular Weight: 254.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 286


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: Benzene-1,2,4,5-tetracarboxylic Acid

Formula: C₁₀H₆O₈
Boiling Pt: 516 °C (1013 hPa)
Melting Pt: 286 °C
Density: 1.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002471
CAS Number: 89-05-4
EINECS: 201-879-5
Merck Index: 12,08188
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2865320308dbae4bb655485dc6d242191399c74&vid=a1d517826c5dfa494e3dc152721ea0e5c8b5199c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR