Benzamidine hydrochloride ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0013-25G B0013-500G B0013-100G
TCB0013-025GEA 61.76 CAD
TCB0013-025G TCB0013-500G TCB0013-100G
Benzamidine hydrochloride ≥96.0% (by titrimetric analysis)
Benzamidine hydrochloride

CAS Number: 1670-14-0
MDL Number: MFCD00013025
Molecular Formula: C7H8N2
Molecular Weight: 156.61
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 171

Formula: C₇H₉ClN₂
Melting Pt: 171 °C
MDL Number: MFCD00066285
CAS Number: 1670-14-0
EINECS: 216-795-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aabcd643dd9da1ea28122459bcc5dddf190d169&vid=1aabcd643dd9da1ea28122459bcc5dddf190d169&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR