3-Amino-1-methyl-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole

A2277-5G A2277-1G
TCA2277-5GEA 74.26 CAD
TCA2277-5G TCA2277-1G
3-Amino-1-methyl-1H-pyrazole ≥98.0%
3-Amino-1-methyl-1H-pyrazole

CAS Number: 1904-31-0
MDL Number: MFCD00466340
Molecular Formula: C4H7N3
Molecular Weight: 97.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 89
Specific Gravity (20/20): 1.12

Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 89 °C
Density: 1.12 (20 °C)
MDL Number: MFCD00466340
CAS Number: 1904-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1c1ba417de2b9bdc044cb4b38f1ba8723170c7&vid=8c1c1ba417de2b9bdc044cb4b38f1ba8723170c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR