2-Amino-4-bromophenol ≥98.0%

Supplier: TCI America

A2271-5G A2271-25G
TCA2271-5GEA 50.56 CAD
TCA2271-5G TCA2271-25G
2-Amino-4-bromophenol ≥98.0%
2-Amino-4-bromophenol

CAS Number: 40925-68-6
MDL Number: MFCD00235171
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: 5-Bromo-2-hydroxyaniline

Formula: C₆H₆BrNO
Melting Pt: 133 °C
MDL Number: MFCD00235171
CAS Number: 40925-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29d518c7c89711a3ee23b49db3351da4bb12fa15&vid=29d518c7c89711a3ee23b49db3351da4bb12fa15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR