2-Amino-6-iodopurine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2068-5G A2068-25G
TCA2068-5GEA 143.78 CAD
TCA2068-5G TCA2068-25G
2-Amino-6-iodopurine ≥98.0% (by HPLC, titration analysis)
2-Amino-6-iodopurine

CAS Number: 19690-23-4
MDL Number: MFCD03840496
Molecular Formula: C5H4IN5
Molecular Weight: 261.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 240

Formula: C₅H₄IN₅
Melting Pt: 240 °C
MDL Number: MFCD08706375
CAS Number: 19690-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2081f8fe26eb603b752ff710d39ee4cba2f66e6&vid=f2081f8fe26eb603b752ff710d39ee4cba2f66e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR