5-Amino-2-chlorophenol ≥98.0%
Supplier: TCI America
Synonyms:
2-Chloro-5-aminophenol
A1976-5G
A1976-25G
TCA1976-5GEA
72.68
CAD
TCA1976-5G
TCA1976-25G
5-Amino-2-chlorophenol ≥98.0%
5-Amino-2-chlorophenol
CAS Number: 6358-06-1
MDL Number: MFCD01707858
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160
Formula:
C₆H₆ClNO Melting Pt: 160 °C |
MDL Number:
MFCD01707858 CAS Number: 6358-06-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...