2-Amino-5-chlorophenol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Chloro-2-hydroxyaniline
A1958-25G
A1958-5G
TCA1958-25GEA
152
CAD
TCA1958-25G
TCA1958-5G
2-Amino-5-chlorophenol ≥97.0% (by GC, titration analysis)
2-Amino-5-chlorophenol
CAS Number: 28443-50-7
MDL Number: MFCD02093863
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 155
Formula:
C₆H₆ClNO MW: 143.57 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD02093863 CAS Number: 28443-50-7 EINECS: 249-020-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...