2-Amino-3,5-dichloro-6-methylpyridine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Amino-3,5-dichloro-2-methylpyridine

A1933-1G A1933-5G
TCA1933-1GEA 26.86 CAD
TCA1933-1G TCA1933-5G
2-Amino-3,5-dichloro-6-methylpyridine ≥97.0% (by titrimetric analysis)
2-Amino-3,5-dichloro-6-methylpyridine

CAS Number: 22137-52-6
MDL Number: MFCD00129029
Molecular Formula: C6H6Cl2N2
Molecular Weight: 177.03
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 6-Amino-3,5-dichloro-2-picoline

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00129029
CAS Number: 22137-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff5b3988e5d2742fbff15ac531a89ac832c38aee&vid=ff5b3988e5d2742fbff15ac531a89ac832c38aee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR