2-Amino-5-chloro-3-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-3-methyl-2-pyridylamine, 5-Chloro-3-methylpyridin-2-amine, 2-Amino-5-chloro-3-picoline

A1931-1G A1931-5G
TCA1931-1GEA 88.48 CAD
TCA1931-1G TCA1931-5G
2-Amino-5-chloro-3-methylpyridine ≥98.0%
2-Amino-5-chloro-3-methylpyridine

CAS Number: 20712-16-7
MDL Number: MFCD00129030
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 70


Other Product Names / Synonyms: 2-Amino-5-chloro-3-picoline

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00129030
CAS Number: 20712-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=587b601ed167c77978f2e0e684ed59d48d54f430&vid=587b601ed167c77978f2e0e684ed59d48d54f430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR