2-Amino-3-chloro-5-methylpyridine ≥98.0%

Supplier: TCI America

A1887-1G A1887-5G
TCA1887-1GEA 67.78 CAD
TCA1887-1G TCA1887-5G
2-Amino-3-chloro-5-methylpyridine ≥98.0%
2-Amino-3-chloro-5-methylpyridine

CAS Number: 31430-41-8
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 60


Other Product Names / Synonyms: 6-Amino-5-chloro-3-picoline

Formula: C₆H₇ClN₂
Melting Pt: 60 °C
MDL Number: MFCD07368858
CAS Number: 31430-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b5f434d11ffcec7d75ad4b996ea71b324e65e8&vid=b1b5f434d11ffcec7d75ad4b996ea71b324e65e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR