2-Amino-m-cresol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1851-25G A1851-5G
TCA1851-25GEA 583.02 CAD
TCA1851-25G TCA1851-5G
2-Amino-m-cresol ≥98.0% (by GC, titration analysis)
2-Amino-m-cresol

CAS Number: 2835-97-4
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: 2-Amino-3-methylphenol, 2-Hydroxy-6-methylaniline

Formula: C₇H₉NO
Melting Pt: 151 °C
MDL Number: MFCD00075082
CAS Number: 2835-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bdf4cbefdb595ae2907e7c8b481c4a9c40f8128&vid=0bdf4cbefdb595ae2907e7c8b481c4a9c40f8128&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR