3-Amino-o-cresol ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-2-methylphenol, 1-Amino-3-hydroxy-2-methylbenzene, 3-Hydroxy-2-methylaniline, 2-Amino-6-hydroxytoluene

A1824-25G A1824-5G
TCA1824-25GEA 595.66 CAD
TCA1824-25G TCA1824-5G
3-Amino-o-cresol ≥98.0%
3-Amino-o-cresol

CAS Number: 53222-92-7
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: 3-Amino-2-methylphenol, 3-Hydroxy-2-methylaniline

Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00007787
CAS Number: 53222-92-7
EINECS: 258-438-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e450f27e370c86f860af9350e7c4df6bd7e6c13d&vid=e450f27e370c86f860af9350e7c4df6bd7e6c13d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR