3-Amino-2,6-dichloropyridine ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine

A1580-25G A1580-5G
TCA1580-25GEA 373.51 CAD
TCA1580-25G TCA1580-5G
3-Amino-2,6-dichloropyridine ≥98.0%
3-Amino-2,6-dichloropyridine

CAS Number: 62476-56-6
MDL Number: MFCD00023417
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 120

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 305
Melting Pt: 120 °C
Density: 1.497 g/cm³ (20 °C)
MDL Number: MFCD00023417
CAS Number: 62476-56-6
EINECS: 263-559-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7fa2aa706aa620c34b51377c8530b3aa4f176da&vid=c7fa2aa706aa620c34b51377c8530b3aa4f176da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR