4-Amino-o-cresol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1536-5G A1536-25G
TCA1536-005GEA 259.44 CAD
TCA1536-005G TCA1536-025G
4-Amino-o-cresol ≥98.0% (by GC, titration analysis)
4-Amino-o-cresol

CAS Number: 2835-96-3
MDL Number: MFCD00007878
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 170


Other Product Names / Synonyms: 4-Amino-2-methylphenol, 4-Hydroxy-3-methylaniline

Formula: C₇H₉NO
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 170 °C
MDL Number: MFCD00007878
CAS Number: 2835-96-3
EINECS: 220-619-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acbf19bc6081173fe8c23a1c89c1a5ed0443fa5d&vid=acbf19bc6081173fe8c23a1c89c1a5ed0443fa5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR