Methyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1499-25G
A1499-5G
TCA1499-025GEA
268.6
CAD
TCA1499-025G
TCA1499-005G
Methyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)
Methyl 3-aminobenzoate
CAS Number: 4518-10-9
MDL Number: MFCD00017102
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 53
Other Product Names / Synonyms: 3-Aminobenzoic Acid Methyl Ester
Formula:
C₈H₉NO₂ Boiling Pt: 170 °C Melting Pt: 53 °C Density: 1.232 g/cm³ (20 °C) |
MDL Number:
MFCD00017102 CAS Number: 4518-10-9 EINECS: 224-842-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...