2-Acetyl-5-chlorothiophene ≥99.0%

Supplier: TCI America

A1471-25G A1471-5G
TCA1471-25GEA 316 CAD
TCA1471-25G TCA1471-005G
2-Acetyl-5-chlorothiophene ≥99.0%
2-Acetyl-5-chlorothiophene

CAS Number: 6310-09-4
MDL Number: MFCD00005444
Molecular Formula: C6H5ClOS
Molecular Weight: 160.62
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 46
Flash Point (°C): 108

Formula: C₆H₅ClOS
Boiling Pt: 118 °C
Melting Pt: 46 °C
Flash Pt: 108 °C
MDL Number: MFCD00005444
CAS Number: 6310-09-4
EINECS: 228-630-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=480be349890f1b331ca30e5962308f7af040ee45&vid=480be349890f1b331ca30e5962308f7af040ee45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR