1-Aminobenzotriazole ≥98.0%

Supplier: TCI America

Synonyms: 1H-Benzo[d][1,2,3]triazol-1-amine

A1464-5G A1464-1G
TCA1464-5GEA 1284.54 CAD
TCA1464-5G TCA1464-001G
1-Aminobenzotriazole ≥98.0%
1-Aminobenzotriazole

CAS Number: 1614-12-6
MDL Number: MFCD00132902
Molecular Formula: C6H6N4
Molecular Weight: 134.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Multi-component Reaction), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-Benzotriazolamine

Formula: C₆H₆N₄
Melting Pt: 84 °C
MDL Number: MFCD00132902
CAS Number: 1614-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=505ba67439bf11a1fa139ac4810cdc6483ec1938&vid=505ba67439bf11a1fa139ac4810cdc6483ec1938&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR