2-Aminopurine ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Purin-2-amine, 7H-Purin-2-amine
A1111-1G
A1111-200MG
TCA1111-001GEA
1538.92
CAD
TCA1111-001G
TCA1111-200MG
2-Aminopurine ≥98.0%
2-Aminopurine
CAS Number: 452-06-2
MDL Number: MFCD00005566
Molecular Formula: C5H5N5
Molecular Weight: 135.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 278
Formula:
C₅H₅N₅ Melting Pt: 278 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005566 CAS Number: 452-06-2 EINECS: 207-197-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...