2-Amino-9-fluorenone ≥98.0% (by HPLC)

Supplier: TCI America

A1040-1G A1040-5G
TCA1040-001GEA 83.74 CAD
TCA1040-001G TCA1040-005G
2-Amino-9-fluorenone ≥98.0% (by HPLC)
2-Amino-9-fluorenone

CAS Number: 3096-57-9
MDL Number: MFCD00001157
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 156

Formula: C₁₃H₉NO
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 156 °C
MDL Number: MFCD00001157
CAS Number: 3096-57-9
EINECS: 221-445-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39352b28259ca900b5edefae93ff31ec41864e44&vid=39352b28259ca900b5edefae93ff31ec41864e44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR