5-Amino-2-methylphenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0925-500G A0925-25G
TCA0925-500GEA 936.94 CAD
TCA0925-500G TCA0925-025G
5-Amino-2-methylphenol ≥98.0% (by titrimetric analysis)
5-Amino-2-methylphenol

Crystals.


Other Product Names / Synonyms: 5-Amino-2-methylphenol, 3-Hydroxy-4-methylaniline

Formula: C₇H₉NO
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 160 °C
MDL Number: MFCD00043922
CAS Number: 2835-95-2
EINECS: 220-618-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af4a5e5a8a2ca38132d65dd5d7aa27bba4bfd1d0&vid=af4a5e5a8a2ca38132d65dd5d7aa27bba4bfd1d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR