1-Amino-2-methylnaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylnaphthalen-1-amine, 2-Methyl-1-naphthylamine

A0917-5G A0917-25G
TCA0917-005GEA 143.78 CAD
TCA0917-005G TCA0917-025G
1-Amino-2-methylnaphthalene ≥98.0%
1-Amino-2-methylnaphthalene

CAS Number: 2246-44-8
MDL Number: MFCD00059135
Molecular Formula: C11H11N
Molecular Weight: 157.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Freezing point (°C): 28
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Methyl-1-naphthylamine

Formula: C₁₁H₁₁N
MW: 157.22 g/mol
MDL Number: MFCD00059135
CAS Number: 2246-44-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efab3ce13b6785b71d5fb51157fe0292dab038e5&vid=efab3ce13b6785b71d5fb51157fe0292dab038e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR