Dodecyl acetate ≥95.0%

Supplier: TCI America

A0902-25ML A0902-500ML
TCA0902-025MLEA 39.5 CAD
TCA0902-025ML TCA0902-500ML
Dodecyl acetate ≥95.0%
Dodecyl acetate

CAS Number: 112-66-3
MDL Number: MFCD00008973
Molecular Formula: C14H28O2
Molecular Weight: 228.38
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 140
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Acetic Acid Lauryl Ester, Acetic Acid Dodecyl Ester, Lauryl Acetate

Formula: C₁₄H₂₈O₂
Boiling Pt: 180 °C
Density: 0.86 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00008973
CAS Number: 112-66-3
EINECS: 203-995-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f06a926c20b99cf6ef1a3b9355428394b7f4abe2&vid=f06a926c20b99cf6ef1a3b9355428394b7f4abe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR