2-Acetylcyclopentanone ≥95.0%

Supplier: TCI America

A0869-25ML A0869-5ML
TCA0869-025MLEA 429.76 CAD
TCA0869-025ML TCA0869-5ML
2-Acetylcyclopentanone ≥95.0%
2-Acetylcyclopentanone

CAS Number: 1670-46-8
MDL Number: MFCD00001413
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 72
Specific Gravity (20/20): 1.06

Formula: C₇H₁₀O₂
Boiling Pt: 103 °C
Density: 1.06 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00001413
CAS Number: 1670-46-8
EINECS: 216-797-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55efbd46bd0db09e217494c9a162772df1fb752f&vid=55efbd46bd0db09e217494c9a162772df1fb752f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR