3-Chloroanthranilic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A0786-25G A0786-5G
TCA0786-25GEA 350.76 CAD
TCA0786-25G TCA0786-5G
3-Chloroanthranilic acid ≥98.0% (by titrimetric analysis)
3-Chloroanthranilic acid

CAS Number: 6388-47-2
MDL Number: MFCD00134229
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 193


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Amino-3-chlorobenzoic Acid

Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 193 °C
MDL Number: MFCD00134229
CAS Number: 6388-47-2
EINECS: 228-996-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=388ddf089fc3554ed24feec53bd0b41c0035da4f&vid=1c0c4949e5ca0a499c6e5c21ce1f61c803aecbb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR