Octadecyl acetate ≥97.0%

Supplier: TCI America

A0675-250G A0675-25G
TCA0675-250GEA 522.98 CAD
TCA0675-250G TCA0675-025G
Octadecyl acetate ≥97.0%
Octadecyl acetate

CAS Number: 822-23-1
MDL Number: MFCD00043649
Molecular Formula: C20H40O2
Molecular Weight: 312.54
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Freezing point (°C): 30


Other Product Names / Synonyms: Acetic Acid Octadecyl Ester, Acetic Acid Stearyl Ester, Stearyl Acetate

Formula: C₂₀H₄₀O₂
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 34.5 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00043649
CAS Number: 822-23-1
EINECS: 212-493-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c65a9e906b574f7fa883455f7192533bade34586&vid=c65a9e906b574f7fa883455f7192533bade34586&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR