2-Aminoquinoline ≥97.0%
Supplier: TCI America
Synonyms:
Quinolin-2-ylamine
A0417-5G
A0417-1G
TCA0417-5GEA
393.42
CAD
TCA0417-5G
TCA0417-001G
2-Aminoquinoline ≥97.0%
2-Aminoquinoline
CAS Number: 580-22-3
MDL Number: MFCD00038079
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 131
Formula:
C₉H₈N₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 131 °C |
MDL Number:
MFCD00038079 CAS Number: 580-22-3 EINECS: 209-458-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...