2-Amino-2-methyl-1,3-propanediol ≥98.0%

Supplier: TCI America

A0332-25G A0332-500G A0332-100G
TCA0332-025GEA 72.68 CAD
TCA0332-025G TCA0332-500G TCA0332-100G
2-Amino-2-methyl-1,3-propanediol ≥98.0%
2-Amino-2-methyl-1,3-propanediol

CAS Number: 115-69-5
MDL Number: MFCD00004678
Molecular Formula: C4H11NO2
Molecular Weight: 105.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 108
Flash Point (°C): 100


Other Product Names / Synonyms: AMPD

Formula: C₄H₁₁NO₂
Boiling Pt: 152 °C
Melting Pt: 108 °C
Density: 1.211 g/cm³ (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
Merck Index: 13,00447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f86c4a3fee6fd26ac8c4c70324660f9ec75cb92c&vid=f86c4a3fee6fd26ac8c4c70324660f9ec75cb92c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR