DL-Homoserine 98,0

Supplier: TCI America

Synonyms: H-DL-HoSer-OH, H-DL-Hser-OH, H-DL-HomoSer-OH, (±)-Homoserine

A0319-5G A0319-1G
TCA0319-5GEA 611.46 CAD
TCA0319-5G TCA0319-001G
DL-Homoserine 98,0
DL-Homoserine

CAS Number: 1927-25-9
MDL Number: MFCD00002618
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 187


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-2-Amino-4-hydroxybutyric Acid

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Boiling Pt: 276
Melting Pt: 151
MDL Number: MFCD00002618
CAS Number: 1927-25-9
EINECS: 217-661-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=486def104a5705b1a2dcbe6a2ec51be1d4bf77c5&vid=486def104a5705b1a2dcbe6a2ec51be1d4bf77c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR