Tyramine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0302-250G A0302-25G A0302-1G
TCA0302-250GEA 1000.14 CAD
TCA0302-250G TCA0302-025G TCA0302-001G
Tyramine ≥98.0% (by GC, titration analysis)
Tyramine

CAS Number: 51-67-2
MDL Number: MFCD00008193
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 207
Melting point (°C): 162


Product Categories: Bioscience, Chemical Mediators, Phenylethylamines
Other Product Names / Synonyms: 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine, 2-(4-Hydroxyphenyl)ethylamine

Formula: C₈H₁₁NO
Boiling Pt: 207 °C
Melting Pt: 162 °C
MDL Number: MFCD00008193
CAS Number: 51-67-2
EINECS: 200-115-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae5d24f9ffe4f1b64869dd67da5ce5d6eb3ce9f3&vid=ae5d24f9ffe4f1b64869dd67da5ce5d6eb3ce9f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR