3-Aminobenzoic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0268-25G A0268-500G
TCA0268-025GEA 281.24 CAD
TCA0268-025G TCA0268-500G
3-Aminobenzoic acid ≥99.0% (by titrimetric analysis)
3-Aminobenzoic acid

CAS Number: 99-05-8
MDL Number: MFCD00007795
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 174
Flash Point (°C): 150

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 174 °C
Density: 1.5105 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4
Merck Index: 13,00420
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8addc1d4f2e8caebecfaeade1987cfa7e07dfa3&vid=d7cd6194ca7d8c7f6bd6b01439ffd9f8f824c2bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR