Adenosine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

A0152-25G A0152-5G A0152-100G
TCA0152-025GEA 105.86 CAD
TCA0152-025G TCA0152-5G TCA0152-100G
Adenosine ≥99.0% (by HPLC, titration analysis)
Adenosine

CAS Number: 58-61-7
MDL Number: MFCD00005752
Molecular Formula: C10H13N5O4
Molecular Weight: 267.25
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: -70 deg (C=2, NaOH sol.(5%))

Formula: C₁₀H₁₃N₅O₄
MW: 267.24 g/mol
Melting Pt: 235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8f98b56cc591e6ddd4e52847b503721638b6a81&vid=a8f98b56cc591e6ddd4e52847b503721638b6a81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR