Nonyl acetate ≥99.0%

Supplier: TCI America

A0041-500ML A0041-25ML
TCA0041-500MLEA 295.46 CAD
TCA0041-500ML TCA0041-025ML
Nonyl acetate ≥99.0%
Nonyl acetate

CAS Number: 143-13-5
MDL Number: MFCD00027340
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 108
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid Nonyl Ester

Formula: C₁₁H₂₂O₂
Boiling Pt: 228 °C
Melting Pt: –26 °C
Density: 0.87 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00027340
CAS Number: 143-13-5
EINECS: 205-585-8
Merck Index: 12,06773
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=582bd41a5e6650c9f32e2a652c1081b3f795e678&vid=582bd41a5e6650c9f32e2a652c1081b3f795e678&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR