2-Bromoethyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic acid (2-bromoethyl) ester

A0023-25G A0023-500G
TCA0023-025GEA 69.52 CAD
TCA0023-025G TCA0023-500G
2-Bromoethyl acetate ≥98.0%
2-Bromoethyl acetate

CAS Number: 927-68-4
MDL Number: MFCD00000235
Molecular Formula: C4H7BrO2
Molecular Weight: 167.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 71
Specific Gravity (20/20): 1.52


Other Product Names / Synonyms: Acetic Acid 2-Bromoethyl Ester

Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 162 °C
Melting Pt: –14 °C
Density: 1.52 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00000235
CAS Number: 927-68-4
EINECS: 213-159-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6772feda2508068160175edb3d3349f91a20131a&vid=6772feda2508068160175edb3d3349f91a20131a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR